Anti-STRADB Goat Polyclonal Antibody
Catalog # PRSI45-246
Supplier: ProSci Inc.
Specifications
- Antigen name:STE20-Related Kinase Adapter Beta
- Antigen symbol:STRADB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Als2cr2|AA792893|PRO1038|ILPIPA|D1Ucla2|ILPIP|B830008M19|Syradb|Papk
- Concentration:500 ug/mL
- Immunogen:ALS2CR2 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of ALS2CR2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ALS2CR2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ALS2CR2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: STRADB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human