Anti-ADH5 Goat Polyclonal Antibody
Catalog # PRSI45-207
Supplier: ProSci Inc.
Specifications
- Antigen name:Alcohol Dehydrogenase 5
- Antigen symbol:ADH5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:glutathione-dependent formaldehyde dehydrogenase|class III alcohol dehydrogenase 5 chi subunit|GSNOR|Alcohol dehydrogenase (class III)|formaldehyde dehydrogenase|alcohol dehydrogenase 5 (class III)|ADH-3|ADH5|FDH |chi polypeptide|ADHX
- Concentration:500 ug/mL
- Immunogen:ADH5 antibody was raised against a 12 amino acid synthetic peptide near the internal region of ADH5.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ADH5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ADH5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ADH5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse