Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ACAP2 Goat Polyclonal Antibody
Anti-ACAP2 Goat Polyclonal Antibody
Catalog # PRSI45-192
Supplier:  ProSci Inc.
undefined
Anti-ACAP2 Goat Polyclonal Antibody
Catalog # PRSI45-192
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    ArfGAP With Coiled-Coil, Ankyrin Repeat And PH Domains 2
  • Antigen symbol:
    ACAP2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    CNT-B2|CENTB2
  • Concentration:
    500 ug/mL
  • Immunogen:
    ACAP2 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of ACAP2.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

ACAP2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ACAP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ACAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human