Anti-BAIAP2 Goat Polyclonal Antibody
PRSI45-017
- Antigen name:BAI1-associated protein 2
- Antigen symbol:BAIAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:FLAF3|brain-specific angiogenesis inhibitor 1-associated protein 2|fas ligand-associated factor 3|BAIAP2|BAP2|BAI1-associated protein 2|insulin receptor substrate p53|IRSP53|insulin receptor substrate p53/p58
- Concentration:500 ug/mL
- Immunogen:BAIAP2 antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of BAIAP2.
- Size:100 µg
- Pk:100 µG
BAIAP2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
BAIAP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: BAIAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human