Order Entry
United Kingdom
ContactUsLinkComponent
Anti-RHCG Goat Polyclonal Antibody
Anti-RHCG Goat Polyclonal Antibody
Catalog # PRSI42-920
Supplier:  ProSci Inc.
undefined
Anti-RHCG Goat Polyclonal Antibody
Catalog # PRSI42-920
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Rh Family C Glycoprotein
  • Antigen symbol:
    RHCG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    PDRC2|SLC42A3|C15orf6|RHGK
  • Concentration:
    500 ug/mL
  • Immunogen:
    Rhcg Antibody was raised against a 11 amino acid sequence near the N-Terminus of Rhcg (mouse).
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Rhcg antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RHCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0