Anti-EFNB2 Goat Polyclonal Antibody
Catalog # PRSI42-823
Supplier: ProSci Inc.
Specifications
- Antigen name:Ephrin B2
- Antigen symbol:EFNB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:EPLG5|HTKL|LERK5|EPH-related receptor tyrosine kinase ligand 5|Ligand of eph-related kinase 5|Htk-L
- Concentration:500 ug/mL
- Immunogen:EFNB2 antibody was raised against a 13 amino acid synthetic peptide near the internal region of EFNB2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
EFNB2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EFNB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat