Anti-IGF1 Goat Polyclonal Antibody
Catalog # PRSI42-659
Supplier: ProSci Inc.
Specifications
- Antigen name:Insulin Like Growth Factor 1
- Antigen symbol:IGF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:Mechano growth factor|Insulin-like growth factor I|IGF-I|MGF|Somatomedin-C|IGFI
- Concentration:500 ug/mL
- Immunogen:IGF1 antibody was raised against a 10 amino acid synthetic peptide near the internal region of IGF1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
IGF1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
IGF1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: IGF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human