Anti-GNAQ Goat Polyclonal Antibody
PRSI42-414
- Antigen name:Guanine Nucleotide Binding Protein (G Protein), Q Polypeptide
- Antigen symbol:GNAQ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:SWS|G-ALPHA-q|GAQ|CMC1
- Concentration:500 ug/mL
- Immunogen:GNAQ antibody was raised against a 14 amino acid synthetic peptide near the internal region of GNAQ (aa 162-175).
- Size:100 µg
- Pk:100 µG
GNAQ antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
GNAQ antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GNAQ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat