Order Entry
United Kingdom
ContactUsLinkComponent
Anti-MXRA5 Goat Polyclonal Antibody
Anti-MXRA5 Goat Polyclonal Antibody
Catalog # PRSI42-243
Supplier:  ProSci Inc.
undefined
Anti-MXRA5 Goat Polyclonal Antibody
Catalog # PRSI42-243
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Matrix-remodelling Associated 5
  • Antigen symbol:
    MXRA5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    Matrix-remodeling-associated protein 5|Adlican|Adhesion protein with leucine-rich repeats and immunoglobulin domains related to perlecan
  • Concentration:
    500 ug/mL
  • Immunogen:
    MXRA5 antibody was raised against a 14 amino acid synthetic peptide near the internal region of MXRA5.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

MXRA5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MXRA5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MXRA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: