Specifications
- Antigen name:Resistin Like Beta
- Antigen symbol:RETNLB
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:XCP2|HXCP2|FIZZ1|RELMb|FIZZ2|RELMbeta|RELM-beta
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mRELMb. Human RELMb; specific antibody was purified by affinity chromatography and then biotinylated.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
RELM beta, is a secreted protein, a probable hormone, expressed only in the gastrointestinal tract, particularly the colon. The biological functions of RELM beta and its molecular targets, are not fully known but, it has been suggested that it plays a regulatory role during inflammation and may also act to establish links among adipose tissue, the intestine and the liver. Interestingly the molecular structure of RELM beta is highly homologous to that of the adipose-derived cytokine Resistin and RELM alpha.
RELMb specific antibody was purified by affinity chromatography and then biotinylated.
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type:
Antigen: RETNLB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human