Anti-FOXG1 Rabbit Polyclonal Antibody
PRSI31-247
- Antigen name:Forkhead Box Protein G1
- Antigen symbol:FOXG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:Brain factor 2|KHL2|HBF-3|QIN|HBF-G2|FOXG1A|HFK1|HFK2|HFK3|FKHL4|Brain factor 1|FKHL3|Forkhead box protein G1|FKH2|BF2|FKHL1|HBF2|FOXG1B|BF1|FOXG1C|HBF-1|FKHL2
- Immunogen:A synthetic peptide corresponding a region of human FOXG1A.
- Size:100 µg
- Pk:100 µG
Forkhead-related protein FKHL2 (brain factor 2, FOXG1A, Forkhead box protein G1A, HFK2 ) is a member of the fork head gene family of transcription factors.
Antibody is supplied as total IgG.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 200 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FOXG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human