Anti-TCF7 Rabbit Polyclonal Antibody
PRSI31-214
- Antigen name:Transcription Factor 7
- Antigen symbol:TCF7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:T-cell specific|TCF7|TCF-1|HMG-box|TCF1|Transcription factor 7
- Immunogen:A synthetic peptide corresponding a region of human TCF7.
- Size:50 µg
- Pk:50 µG
The T-cell-specific transcription factor TCF7 activates genes involved in immune regulation and is a candidate locus for genetic susceptibility to type 1 diabetes.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TCF7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human