Anti-ZNF33A Rabbit Polyclonal Antibody
PRSI30-732
- Antigen name:Zinc Finger Protein 33A
- Antigen symbol:ZNF33A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:ZNF11|KOX31|zinc finger protein 33A|ZNF33|NF11A|ZZAPK|KOX2|KOX5|ZNF11A
- Immunogen:A synthetic peptide corresponding a region of human ZNF33A.
- Size:50 µg
- Pk:50 µG
ZNF33A belongs to the krueppel C2H2-type zinc-finger protein family. It contains 16 C2H2-type zinc fingers and 1 KRAB domain. ZNF33A may be involved in transcriptional regulation.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZNF33A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human