Anti-APC2 Rabbit Polyclonal Antibody
Catalog # PRSI30-627
Supplier: ProSci Inc.
Specifications
- Antigen name:Adenomatosis Polyposis Coli 2
- Antigen symbol:APC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:anaphase-promoting complex subunit 2|APCL|cyclosome subunit 2
- Immunogen:A synthetic peptide corresponding a region of human APC2.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
APC2 belongs to the adenomatous polyposis coli (APC) family. It contains 6 ARM repeats. APC2 promotes rapid degradation of CTNNB1 and may function as a tumor suppressor. It may function in Wnt signaling.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: APC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat