Anti-STEAP4 Rabbit Polyclonal Antibody
PRSI30-549
- Antigen name:Six Transmembrane Epithelial Antigen of the Prostate 4
- Antigen symbol:STEAP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:TIARP |tumor necrosis factor|tumor necrosis-alpha-induced adipose-related protein|six transmembrane prostate protein 2 |STAMP2 |alpha-induced protein 9 |FLJ23153 |DKFZp666D049 |STEAP family member 4|TNFAIP9
- Immunogen:A synthetic peptide corresponding a region of human STEAP4.
- Size:50 µg
- Pk:50 µG
Metalloreductase has the ability to reduce both Fe (3+) to Fe (2+) and Cu (2+) to Cu (1+). It uses NAD (+) as acceptor.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: STEAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human