Anti-GCNT4 Rabbit Polyclonal Antibody
PRSI30-275
- Antigen name:Glucosaminyl (N-acetyl) Transferase 4
- Antigen symbol:GCNT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Dog
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:C2GNT3
- Immunogen:A synthetic peptide corresponding a region of human GCNT4.
- Size:50 µg
- Pk:50 µG
GCNT4 is a glycosyltransferase that mediates core 2 O-glycan branching, an important step in mucin-type biosynthesis. It does not have core 4 O-glycan or I-branching enzyme activity.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GCNT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog