Anti-SLC39A12 Rabbit Polyclonal Antibody
Catalog # PRSI29-953
Supplier: ProSci Inc.
Specifications
- Antigen name:Solute Carrier Family 39 (Zinc transporter) Member 12
- Antigen symbol:SLC39A12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:LZT-Hs8|ZIP12|bA570F3.1|Zinc transporter ZIP12|ZIP-12|LIV-1 subfamily of ZIP zinc transporter 8
- Immunogen:A synthetic peptide corresponding a region of human SLC39A12.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC39A12 may act as a zinc-influx transporter and also may be partly involved in the outbreak of schizophrenia.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC39A12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human