Anti-PNPT1 Rabbit Polyclonal Antibody
Catalog # PRSI29-484
Supplier: ProSci Inc.
Specifications
- Antigen name:Polyribonucleotide Nucleotidyltransferase 1
- Antigen symbol:PNPT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:PNPASE|old-35|COXPD13|DFNB70|OLD35
- Immunogen:A synthetic peptide corresponding a region of human PNPT1.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
PNPT1 is a subunit of the exosome complex, which is involved in 3-prime-to-5-prime exoribonuclease activity for RNA processing and degradation.PNPT1 is a subunit of the exosome complex, which is involved in 3-prime-to-5-prime exoribonuclease activity for RNA processing and degradation (Raijmakers et al., 2002 [PubMed 12419256]).
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PNPT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human