Anti-NOVA2 Rabbit Polyclonal Antibody
PRSI29-290
- Antigen name:Neuro-Oncological ventral Antigen 2
- Antigen symbol:NOVA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:ANOVA|NOVA2_HUMAN.|Astrocytic NOVA1 like RNA binding protein|Gm1424
- Immunogen:A synthetic peptide corresponding a region of human NOVA2.
- Size:100 µg
- Pk:100 µG
NOVA2 may regulate RNA splicing or metabolism in a specific subset of developing neurons. It binds single strand RNA.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NOVA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat