Specifications
- Antigen name:Thyroid Hormone Receptor Interactor 13
- Antigen symbol:TRIP13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:Human papillomavirus type 16 E1 protein-binding protein|Thyroid hormone receptor interactor 13|TRIP13|16E1-BP|HPV16 E1 protein-binding protein|TRIP-13|Thyroid receptor-interacting protein 13
- Immunogen:A synthetic peptide corresponding a region of human TRIP13.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The thyroid hormone (T3) receptors (TRs) are hormone-dependent transcription factors that regulate expression of a variety of specific target genes. TRIP13 specifically interacts with the ligand binding domain of the TRs. It is a member of Trips (TR-interacting proteins) family. Nearly all of the Trips also show similar ligand-dependent interaction with the retinoid X receptor (RXR), but none interact with the glucocorticoid receptor under any conditions. Trips predict specific functional roles: one is an apparent human homolog of a yeast transcriptional coactivator, one is a new member of a class of nonhistone chromosomal proteins, and one contains a conserved domain associated with ubiquitination of specific target proteins.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRIP13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human