Anti-HMG20B Rabbit Polyclonal Antibody
Catalog # PRSI28-611
Supplier: ProSci Inc.
Specifications
- Antigen name:High Mobility Group 20B
- Antigen symbol:HMG20B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:BRAF25|SOXL|pp8857|SMARCE1r|PP7706|BRAF35|HMGXB2|HMGX2
- Immunogen:A synthetic peptide corresponding a region of mouse HMG20B.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Hmg20b is the component of a BHC histone deacetylase complex that contains HDAC1, HDAC2, HMG20B/BRAF35, AOF2/LSD1, RCOR1/CoREST and PHF21A/BHC80. Hmg20b is required for correct progression through G2 phase of the cell cycle and entry into mitosis Hmg20b is also required for RCOR1/CoREST mediated repression of neuronal specific gene promoters.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: HMG20B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat