Anti-NKX2-3 Rabbit Polyclonal Antibody
PRSI28-591
- Antigen name:Nk2 Homeobox 3
- Antigen symbol:NKX2-3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:CSX3|NK2.3|NKX2C|NKX4-3|NKX2.3
- Immunogen:A synthetic peptide corresponding a region of human NKX2-3.
- Size:100 µg
- Pk:100 µG
NKX2C is a member of the NKX family of homeodomain-containing transcription factors, which are implicated in many aspects of cell type specification and maintenance of differentiated tissue functions.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NKX2-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat