Specifications
- Antigen name:Thyroid Hormone Receptor Interactor 4
- Antigen symbol:TRIP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Dog,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:ASC-1|ASC1|HsT17391|ZC2HC5
- Immunogen:A synthetic peptide corresponding a region of human TRIP4.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
TRIP4 is a transcription coactivator of nuclear receptors which functions in conjunction with CBP-p300 and SRC-1 and may play an important role in establishing distinct coactivator complexes under different cellular conditions. TRIP4 plays a pivotal role in the transactivation of NF-kappa-B, SRF and AP1. TRIP4 acts as a mediator of transrepression between nuclear receptor and either AP1 or NF-kappa-B. It plays a role in androgen receptor transactivation and in testicular function
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRIP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse