Anti-NMUR2 Rabbit Polyclonal Antibody
PRSI28-267
- Antigen name:Neuromedin U Receptor 2
- Antigen symbol:NMUR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:FM-4|TGR-1|NMU-R2|TGR1|NMU2R|FM4
- Immunogen:A synthetic peptide corresponding a region of human NMUR2.
- Size:100 µg
- Pk:100 µG
NMUR2 encodes for one of two G-protein-coupled receptors for the neuropeptide, neuromedin U. This peptide is found in highest levels in the gut and genitourinary system where it potently contracts smooth muscle but is also expressed in the spinal cord and discrete regions of the brain. NMUR2 is highly expressed in the central nervous system.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NMUR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human