Specifications
- Antigen name:Transcription Factor B1 Mitochondrial
- Antigen symbol:TFB1M
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:homolog of yeast mitochondrial transcription factor B|N'-adenosyl(rRNA) dimethyltransferase 1|mitochondrial 12S rRNA dimethylase 1|S-adenosylmethionine-6-N'|mtTFB|CGI-75|mitochondrial|CGI75|transcription factor B1|OTTHUMP00000017473|mtTFB1|TFB1M
- Immunogen:A synthetic peptide corresponding a region of human TFB1M.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The transcription of genes from mitochondrial DNA requires a mitochondrial RNA polymerase and a DNA-binding transcription factor. Transcription factor B1 (TFB1M) is a part of this transcription complex.The transcription of genes from mitochondrial DNA requires a mitochondrial RNA polymerase (see POLRMT, MIM 601778) and a DNA-binding transcription factor (see TFAM, MIM 600438). Transcription factor B1 (TFB1M) is a part of this transcription complex.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TFB1M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human