- Antigen name:Zinc Finger And SCAN Domain Containing 25
- Antigen symbol:ZSCAN25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human ZNF498.
- Size:100 µg
- Pk:100 µG
The function of ZNF498 remains unknown. The protein bears some similarity to zinc finger proteins, which are involved in DNA binding and protein-protein interactions. Alternative splicing results in two transcript variants encoding different proteins. Additional splice variants have been identified, but their biological validity has not been determinedThis gene encodes a protein of unknown function. The protein bears some similarity to zinc finger proteins, which are involved in DNA binding and protein-protein interactions.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ZSCAN25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human