Anti-PCGF3 Rabbit Polyclonal Antibody
Catalog # PRSI28-083
Supplier: ProSci Inc.
Specifications
- Antigen name:Polycomb group ring Finger 3
- Antigen symbol:PCGF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:PCGF3_HUMAN.|polycomb group ring finger 3|DONG1|PCGF3|Ring finger protein 3|RNF3A|RNF3
- Immunogen:A synthetic peptide corresponding a region of human PCGF3.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
PCGF3 contains a C3HC4 type RING finger, which is a motif known to be involved in protein-protein interactions. The specific function of this protein has not yet been determined.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PCGF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human