Order Entry
United Kingdom
ContactUsLinkComponent
Anti-GTF2H3 Rabbit Polyclonal Antibody
Anti-GTF2H3 Rabbit Polyclonal Antibody
Catalog # PRSI27-346
Supplier:  ProSci Inc.
undefined
Anti-GTF2H3 Rabbit Polyclonal Antibody
Catalog # PRSI27-346
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    General Transcription Factor 2H Subunit 3
  • Antigen symbol:
    GTF2H3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Lyophilized
  • Antigen synonyms:
    BTF2|TFB4|P34|TFIIH
  • Immunogen:
    A synthetic peptide corresponding a region of human GTF2H3.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

GTranscription Factor Antibodies2H3 interacts with HIV-1 Tat as a component of the HIV-1 transcription pre-initiation complex, but is released from the elongation complex which includes P-TEFb. It synergizes with HIV-1 Tat to induce transcription elongation from the HIV-1 LTR promoter.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GTF2H3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human