Anti-KLHL15 Rabbit Polyclonal Antibody
Catalog # PRSI27-174
Supplier: ProSci Inc.
Specifications
- Antigen name:Kelch-like 15
- Antigen symbol:KLHL15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human KLHL15.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
KLHL15 is a member of the kelch-like family of proteins that share a common domain structure consisting of an N-terminal broad-complex, tramtrack, bric-a-brac/poxvirus and zinc finger domain and C-terminal kelch repeat motifs. KLHL15 may be involved in protein ubiquitination and cytoskeletal organization.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: KLHL15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat