Anti-FHL3 Rabbit Polyclonal Antibody
PRSI26-538
- Antigen name:Four and a Half LIM Domains 3
- Antigen symbol:FHL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:SLIM2
- Immunogen:A synthetic peptide corresponding a region of human FHL3.
- Size:50 µg
- Pk:50 µG
FHL3 contains 4 LIM zinc-binding domains. It is expressed only in skeletal muscle. FHL3 association with MyoD may contribute to the regulation of MyoD-dependent transcription of muscle genes and thereby myogenesis.LIM proteins are defined by the possession of a highly conserved double zinc finger motif called the LIM domain.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FHL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human