- Antigen name:BCL2-related ovarian killer
- Antigen symbol:BOK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:Bcl-2-like protein 9|Bcl-2-related ovarian killer protein|BCL2L9|BOKL|hBOK
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:BOK antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human BOK.
- Tested applications:E, IHC
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
BOK Antibody: Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. The Bcl-2 family of proteins is comprised of critical regulators of apoptosis that can be divided into two classes: those that inhibit apoptosis and those that promote cell death. BOK, a pro-apoptotic Bcl-2 family member, was initially identified in the ovary, and was found to interact with other Bcl-2 family members such as Mcl-1 and Bfl-1. BOK expression is high during early placental development, suggesting that it may also play a role in regulating trophoblast cell proliferation.
Type:
Antigen: BOK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human