Reference standards, biodiesel/diesel fuel Blends
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
These standards are intended for use as calibration and reference standards for the determination of fatty acid methyl esters (FAME) biodiesel content in diesel fuel by infrared (IR) spectroscopy. They are formulated specifically for use with ASTM Method D7371 and EN 14078.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together