Order Entry
United Kingdom
ContactUsLinkComponent
Drug and metabolite standards, clobenzorex hydrochloride 1,0 mg/ml in methanol, LoGiCal®
Drug and metabolite standards, clobenzorex hydrochloride 1,0 mg/ml in methanol, LoGiCal®
  PROOAMP0649.00-01
 :  
Drug and metabolite standards, clobenzorex hydrochloride 1,0 mg/ml in methanol, LoGiCal®
  PROOAMP0649.00-01
 :  

 

  • Pk:
    1 mL
  • Standards:
    Reference standard
  • CAS No.:
    5843-53-8
  • Description:
    Clobenzorex hydrochloride 1,0 mg/ml in methanol (as free base)
  • Environmentally Preferable:
  • Formula:
    C₁₆H₁₉Cl₂N
  • Description:
    Clobenzorex hydrochloride