Containment berms, QuickBerm® Lite
Supplier: JUSTRITE MANUFACTURING
Certificates
About this item
Portable, versatile containment berm deploys quickly for emergency spill response.
- Leakproof, tough Durable EnGuard™I, PVC coated fabric provides broad UV and chemical resistance that withstands repeated usage
- Reinforced inside supports reduce sagging and prevent tripping hazards that can be caused by outside supports
Lightweight berm folds down easily to a small size for storage in trucks, emergency vehicles, and spill kits. Add optional grates to help reduce contact with contaminated liquids by personnel or equipment.
Certifications: Helps to comply with EPA for containment and spill prevention (SPCC).
Frequently Bought Together