Electric barrel pump
Catalog # BURK5780-0040
Supplier: BURKLE
Specifications
- Flow rate:40 l/min
- Description:Electric barrel pump petro, standard
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Electric barrel pump for pumping diesel oil, Category EL fuel oil, undiluted radiator antifreeze and other liquids of Hazard Category AIII (VbF).
Built-in siphon protection prevents leaks if the tube is faulty. Available for 230 V.
- With manual suction pump
- VDE tested, motor with thermal protection switch
- Barrel thread M 64×4 and 2"
- Suction hose with filter 1,60 m length
- Pump hose made of Perbunan, 4 m length, with bend protection spring and pump nozzle
- Not suitable for aqueous/diluted liquids