Bioreactors, UniVessel® SU
Catalog # SARPDUO002LL-SS01V
Supplier: Sartorius
Specifications
- Description:UniVessel® SU single-use bioreactor
- Environmentally Preferable:
- Pk:2
Specifications
About this item
The UniVessel® SU is a stirred tank, single-use bioreactor with a working volume range of 0,6 to 2 L. It is assembled, irradiated and shipped ready-to-use.
- Proven and scaleable design
- Compatible with your existing bioreactor controller
- Interchangeable with existing glass vessels
UniVessel® SU is compatible with your controller and can be used interchangeably with glass vessels to help you efficiently manage peak workloads despite challenging timelines.