Cover slips for microscope slides
Supplier: Thermo Fisher Scientific
Certificates
About this item
These cover slips made of fused quartz, have many properties, due to the fusing of naturally occuring crystalline silica.
- High chemical purity and corrosion resistance
- Low thermal expansion coefficient
- Extreme hardness
Excellent electrical insulation and optical transmission from ultraviolet to infrared are suitable for analytical purposes.
Frequently Bought Together