Decontamination solution, DNA AWAY®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Ready-to-use solution for eliminating DNA contamination. For cleaning work surfaces, instruments, pipettes, thermal cyclers and more.
- Suitable for PCR applications, preventing false-positive PCR results
- Easy cleaning of various types of surfaces
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications