VWR® B'Wipes 1000, Cleanroom wipes
Supplier: VWR Collection
Certificates
About this item
Absorbent, non woven wipe for use in ISO 6 cleanrooms.
- Non woven wipe, made of hydroentangled 55% cellulose and 45% polyester (68 g/m²)
- High absorbency, strong and durable
- Works well with IPA and other solvents
- Ideal for spill control and general purpose wiping
Frequently Bought Together