VWR®, Silane Coated, Colour Frosted Slides
Silane coated (aminoalkylsilane) glass slides producing a sticky surface, which can bind tissue sections to glass.
- Smooth edges for operational safety
- Marking with pencils, marker pens and conventional labelling systems
- Shelf life of 18 months
Recommended for H&E stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopathology examination.
: 50 pieces per box. 50 boxes per case.
: Fragile items should be stored in dry and proper laboratory temperature conditions. Not tested for use in microarray assays.
Frequently Bought Together