Anti-HepPar-1 Mouse Monoclonal Antibody [clone: HepPar1]
ABNOMAB14640
: Abnova
- Antibody type:Primary
- Antigen symbol:HepPar-1
- Clonality:Monoclonal
- Clone:HepPar1
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Storage buffer:In 10 mM PBS.
- Storage temperature:Store at -20 to –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Extract of a formalin-fixed, rejected-allograft of a human liver.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Mouse monoclonal antibody raised against human HepPar-1.
Type: Primary
Antigen: HepPar-1
Clonality: Monoclonal
Clone: HepPar1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human