Multi-channel pipettes, manual, variable volume, with LTS™ and RFID tag, Pipet-Lite™ XLS+
Supplier: RAININ
Certificates
About this item
This ergonomic manual pipette with comfortable handle ensure light and smooth operation and significantly reduce the risk of repetitive strain injuries.
- LTS (LiteTouch System) reduces tip ejection forces by 85% and prevents RSI injuries
- Effortless pipetting due to light weight, light plunger forces and lowest tip ejection forces
- Sealing system minimises 'piston stickiness', providing greater control, particularly with applications that require slow, careful pipetting
- Sophisticated mechanical designs (new push-rod design) ensure high performance, and superior channel-to-channel consistency
- On-board RFID provides ease of service and calibration tracking
- Intended for use with LTS™ LiteTouch™ tips
Frequently Bought Together