Pig trypsin (from pancreas), MP Biomedicals
Catalog # ICNA091689149
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:100 mL
- CAS No.:9002-07-7
- Source:Pancreas
- Species:Pig
- Environmentally Preferable:
- Storage conditions:−20 °C
- Form:Pale to dark pink Solution
- Enzyme name:Trypsin
- Molecular weight:23000
- Concentration:0,25% (w/v) trypsin (1:250) and 0,02% (w/v) EDTA
- pH:7,0 - 7,4
Specifications
About this item
Trypsin-EDTA solution is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation due to its digestive strength.
- Sterile
- Concentration: 0.05% (w/v) Trypsin (1:250) and 0.02% (w/v) EDTA (Versene)
- Formulation: EDTA 2Na 2H2O: 200.00000 mg/L, Potassium Chloride [KCl]: 400.00000 mg/L, Sodium Bicarbonate [NaHCO3]: 580.00000 mg/L, Sodium Chloride [NaCl]: 8000.00000 mg/L, Dextrose: 1000.00000 mg/L, Phenol Red Sodium Salt: 2.00000 mg/L, Trypsin 1:250: 500.00000 mg/L
- pH: 7,0 to 7,4
- Foreign Activity: Mycoplasma, Porcine Parvovirus : Not detected
- Osmolality: 270 to 305 mM/kg
- Storage and Handling: −20 °C
Trypsin-EDTA solution is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation due to its digestive strength.