Specifications
- Antibody type:Primary
- Antigen name:fidgetin-like 1
- Antigen symbol:FIGNL1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:63979
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
FIGNL1 is a 674 amino acid protein belonging to the AAA ATPase family. FIGNL1 exists as a hexamer that undergoes alternative splicing to produce two isoforms. FIGNL1 utilizes magnesium as a cofactor and is phosphorylated upon DNA damage, probably by ATM or ATR. FIGNL1 is suggested to regulate osteoblast proliferation and differentiation. FIGNL1 is encoded by a gene located on human chromosome 7, which consists about 158 milllion bases, encodes over 1000 genes and makes up about 5% of the human genome. Chromosome 7 has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterized by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance. Deletions of portions of the q arm of chromosome 7 are also seen in a number of myeloid disorders including cases of acute myelogenous leukemia and myelodysplasia.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FIGNL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat