Anti-SPECC1L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-7773R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Sperm Antigen With Calponin Homology And Coiled-Coil Domains 1-Like
- Antigen symbol:SPECC1L
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:23384
- Antigen synonyms:SPECC1-like protein|Renal carcinoma antigen NY-REN-22|specc1l|CYTSA|CYTSA_HUMAN|Cytokinesis and spindle organization A|Cytospin-A|Sperm antigen with calponin homology and coiled-coil domains 1-like.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Involved in cytokinesis and spindle organization.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SPECC1L
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit