Anti-ITIH1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-7018R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Inter-alpha (globulin) Inhibitor H1
- Antigen symbol:ITIH1
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:3697
- Antigen synonyms:ITI-HC1|Inter-alpha-trypsin inhibitor heavy chain H1|IGHEP1|ITIH1|SHAP|Serum-derived hyaluronan-associated protein|ITI heavy chain H1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
May act as a carrier of hyaluronan in serum or as a binding protein between hyaluronan and other matrix protein, including those on cell surfaces in tissues to regulate the localization, synthesis and degradation of hyaluronan which are essential to cells undergoing biological processes.Contains a potential peptide which could stimulate a broad spectrum of phagocytotic cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ITIH1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat