Anti-TGC Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-6690R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Transglutaminase
- Antigen symbol:TGC
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:7051
- Antigen synonyms:Tissue transglutaminase|TGase-H|TGC|TG(C)|Transglutaminase-2|TGase C
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:500-550/817
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Catalyzes the cross-linking of proteins and the conjugation of polyamines to proteins. Responsible for cross-linking epidermal proteins during formation of the stratum corneum.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Transglutaminase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat