Anti-MMTN Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-6551R-A488
: Bioss
- Antibody type:Primary
- Antigen name:Mimitin
- Antigen symbol:MMTN
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:B17.2 like|B17.2-like|Myc induced mitochondrial protein|Mimitin|NADH dehydrogenase ubiquinone 1 alpha subcomplex assembly factor 2|Myc-induced mitochondrial protein|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2|B17.2L|MIMIT_HUMAN|mitochondrial|NDUFA12 like protein|MMTN|NDUFA12-like protein|Mimitin mitochondrial|NDUFA12 like|NDUFA12L|NDUFAF2.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
NADH:ubiquinone oxidoreductase (complex I) catalyzes the transfer of electrons from NADH to ubiquinone (coenzyme Q) in the first step of the mitochondrial respiratory chain, resulting in the translocation of protons across the inner mitochondrial membrane. This gene encodes a complex I assembly factor. Mutations in this gene cause progressive encephalopathy resulting from mitochondrial complex I deficiency.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MMTN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat