Specifications
- Antibody type:Primary
- Antigen name:DCN1, defective in cullin neddylation 1, Domain Containing 1
- Antigen symbol:DCUN1D1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:54165
- Antigen synonyms:DCN1|domain containing 1 (S. cerevisiae)|DCUN1D1|RP42|SCRO|defective in cullin neddylation 1|squamous cell carcinoma-related oncogene|Tes3|DCUN1L1|SCCRO|RP42 homolog
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
DCUN1D1 (DCUN1 domain-containing protein 1) contains a DUF298 domain and a UBA-like ubiquitin. Forms part of an E3 ubiquitin ligase complex for neddylation. Required for neddylation of cullin components of E3 cullin-RING ubiquitin ligase complexes by enhancing the rate of cullins neddylation. Functions to recruit the NEDD8-charged E2 enzyme to the cullin component. Involved in the release of inhibitory effets of CAND1 on cullin-RING ligase E3 complex assembly and activity. Acts also as an oncogene facilitating malignant transformation and carcinogenic progression. Defects in DCUN1D1 may be a cause of squamous cell carcinomas.Strongly overexpressed in thyroid tumors, bronchioloalveolar carcinomas, and malignant tissues of squamous cell carcinoma of the oral tongue. Not overexpressed in aggressive adrenocortical carcinomas.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DCUN1D1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat