Anti-CPE Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-6478R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:carboxypeptidase E
- Antigen symbol:CPE
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:1363
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:200-250/476
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Removes residual C-terminal Arg or Lys remaining after initial endoprotease cleavage during prohormone processing. Processes proinsulin.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CPE
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity: